3-AP ≥95%
Bottle
Katalog # APOSBUP13017-500MG
New Product
Leverandør: Apollo Scientific
3-AP ≥95%
Katalog # APOSBUP13017-500MG
Leverandør: Apollo Scientific
New Product
CAS nummer: 143621-35-6
Gå til:
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonymer: 3-AP, NSC 663249, 2-((3-Aminopyridin-2-yl)methylene)hydrazine-1-carbothioamide, 2-[(3-Aminopyridin-2-yl)methylene]hydrazinecarbothioamide, PAN-811, 3-Aminopyridine-2-Carboxyaldehyde Thiosemicarbazone
- CAS nummer:143621-35-6
- Formel:C₇H₉N₅S
- Molecular Weight:195,24 g/mol
Specifikationer
Specifikationer
- Pakning:Bottle



