3-AP ≥95%
Bottle
Catalog # APOSBUP13017-100MG
New Product
Supplier: Apollo Scientific
3-AP ≥95%
Catalog # APOSBUP13017-100MG
Supplier: Apollo Scientific
New Product
CAS Number: 143621-35-6
Jump to:
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonyms: 3-AP, NSC 663249, 2-((3-Aminopyridin-2-yl)methylene)hydrazine-1-carbothioamide, 2-[(3-Aminopyridin-2-yl)methylene]hydrazinecarbothioamide, PAN-811, 3-Aminopyridine-2-Carboxyaldehyde Thiosemicarbazone
- CAS Number:143621-35-6
- Formula:C₇H₉N₅S
- Molecular Weight:195,24 g/mol
Specifications
Specifications
- Pakning:Bottle



