Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
3-AP ≥95%

3-AP ≥95%

Bottle
Katalog # APOSBUP13017-25MG
New Product
Leverandør:   Apollo Scientific
3-AP ≥95%
Katalog # APOSBUP13017-25MG
Leverandør:  Apollo Scientific
New Product
CAS nummer:  143621-35-6
3-AP ≥95%

Bottle

CAS nummer:  143621-35-6
Price
1.490,00 kr
Log ind for tilgængelighed
1 * 25 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonymer:  3-AP, NSC 663249, 2-((3-Aminopyridin-2-yl)methylene)hydrazine-1-carbothioamide, 2-[(3-Aminopyridin-2-yl)methylene]hydrazinecarbothioamide, PAN-811, 3-Aminopyridine-2-Carboxyaldehyde Thiosemicarbazone
  • CAS nummer:143621-35-6
  • Formel:C₇H₉N₅S
  • Molecular Weight:195,24 g/mol
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.