4'-Demethylepipodophyllotoxin
Bottle
Katalog # APOSOR3101T-250MG
New Product
Leverandør: Apollo Scientific
4'-Demethylepipodophyllotoxin
Katalog # APOSOR3101T-250MG
Leverandør: Apollo Scientific
New Product
CAS nummer: 6559-91-7
Gå til:
Product Details & Documents
A potent inhibitor of microtubule assembly.
Synonymer: (5R,5aR,8aR,9S)-9-Hydroxy-5-(4-hydroxy-3,5-dimethoxyphenyl)-5,5a,8a,9-tetrahydrofuro[3',4':6,7]naphtho[2,3-d][1,3]dioxol-6(8H)-one
- CAS nummer:6559-91-7
- Formel:C₂₁H₂₀O₈
- MDL nummer:MFCD00189421
- Molecular Weight:400.38 g/mol
- Opbevaringstemperatur:Stue temperatur
Specifikationer
Specifikationer
- Pakning:Bottle
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



