Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
2-Nitrocinnamaldehyde

2-Nitrocinnamaldehyde

Bottle
Katalog # APOSOR28735-500G
Irriterende
New Product
Leverandør:   Apollo Scientific
2-Nitrocinnamaldehyde
Katalog # APOSOR28735-500G
Leverandør:  Apollo Scientific
New Product
CAS nummer:  1466-88-2
2-Nitrocinnamaldehyde

Bottle

CAS nummer:  1466-88-2
Price
20.220,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents
Synonymer:  2'-Nitrocinnamaldehyde
  • CAS nummer:1466-88-2
  • Formel:C₉H₇NO₃
  • MDL nummer:MFCD00007188
  • Molecular Weight:177.16 g/mol
  • Smeltende Pt:127…129 °C
  • Opbevaringstemperatur:Køleskab
  • EINECS:215-988-0
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.