Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Diethylenglycol 99%

Diethylenglycol 99%

Bottle
Katalog # APOSOR59808-250G
Irriterende
Sundhed
New Product
Leverandør:   Apollo Scientific
Diethylenglycol 99%
Katalog # APOSOR59808-250G
Leverandør:  Apollo Scientific
New Product
CAS nummer:  111-46-6
Diethylenglycol 99%

Bottle

CAS nummer:  111-46-6
Price
374,00 kr
Log ind for tilgængelighed
1 * 250 g
Product Details & Documents
Synonymer:  2,2'-Oxydiethanol
  • CAS nummer:111-46-6
  • Formel:C₄H₁₀O₃
  • MDL nummer:MFCD00002882
  • Molecular Weight:106.12 g/mol
  • Kogende Pt:245…246 °C
  • Smeltende Pt:–10 °C
  • Tæthed:1,118 g/cm³ (25 °C)
  • Flash Pt:123 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-872-2
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.