Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Cyclohexanone 2,4-Dinitrophenylhydrazone 99%

Cyclohexanone 2,4-Dinitrophenylhydrazone 99%

Bottle
Katalog # APOSOR1027545-250M
New Product
Leverandør:   Apollo Scientific
Cyclohexanone 2,4-Dinitrophenylhydrazone 99%
Katalog # APOSOR1027545-250M
Leverandør:  Apollo Scientific
New Product
CAS nummer:  1589-62-4
Cyclohexanone 2,4-Dinitrophenylhydrazone 99%

Bottle

CAS nummer:  1589-62-4
Price
418,00 kr
Log ind for tilgængelighed
1 * 250 mg
Product Details & Documents

Usage: Organic building block/reagent

  • CAS nummer:1589-62-4
  • Formel:C₁₂H₁₄N₄O₄
  • Molecular Weight:278,26 g/mol
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.