AC-55541 98%
Bottle
Katalog # APOSOR1026543-1G
New Product
Leverandør: Apollo Scientific
AC-55541 98%
Katalog # APOSOR1026543-1G
Leverandør: Apollo Scientific
New Product
CAS nummer: 916170-19-9
Gå til:
Product Details & Documents
Usage: Organic building block/reagent
Synonymer: (E)-N-(2-(2-(1-(3-Bromophenyl)ethylidene)hydrazineyl)-2-oxo-1-(4-oxo-3,4-dihydrophthalazin-1-yl)ethyl)benzamide
- CAS nummer:916170-19-9
- Formel:C₂₅H₂₀BrN₅O₃
- Molecular Weight:518,36 g/mol
Specifikationer
Specifikationer
- Pakning:Bottle



