Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

2'-Hydroxyflavanone 98%

Bottle
Catalog # APOSOR1029230-25G
New Product
Supplier:   Apollo Scientific
2'-Hydroxyflavanone 98%
Catalog # APOSOR1029230-25G
Supplier:  Apollo Scientific
New Product
CAS Number:  17348-76-4
2'-Hydroxyflavanone 98%

Bottle

CAS Number:  17348-76-4
Price
9.610,00 kr
Log in for availability
1 * 25 g
Product Details & Documents

Usage: Organic building block/reagent.

  • CAS Number:17348-76-4
  • Formula:C₁₅H₁₂O₃
  • Molecular Weight:240,26 g/mol
Specifications

Specifications

  • Pakning:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.