2'-Hydroxyflavanone 98%
Bottle
Katalog # APOSOR1029230-1G
New Product
Leverandør: Apollo Scientific
2'-Hydroxyflavanone 98%
Katalog # APOSOR1029230-1G
Leverandør: Apollo Scientific
New Product
CAS nummer: 17348-76-4
Gå til:
Product Details & Documents
Usage: Organic building block/reagent.
- CAS nummer:17348-76-4
- Formel:C₁₅H₁₂O₃
- Molecular Weight:240,26 g/mol
Specifikationer
Specifikationer
- Pakning:Bottle



