Orders LinkContactUsLinkComponent
2'-Hydroxyflavanone 98%
Bottle
  APOSOR1029230-100M
New Product
2'-Hydroxyflavanone 98%
  APOSOR1029230-100M
New Product
 :  17348-76-4
2'-Hydroxyflavanone 98%
Bottle
 :  17348-76-4
165,00 kr
1 * 100 mg
Usage: Organic building block/reagent.
  •  :17348-76-4
  •  :C₁₅H₁₂O₃
  •  :240,26 g/mol
  • Pakning:
    Bottle