Order Entry
Denmark
Orders LinkContactUsLinkComponent
Gemfibrozil 98%

Gemfibrozil 98%

Bottle
Katalog # APOSOR1029081-2.5K
New Product
Leverandør:   Apollo Scientific
Gemfibrozil 98%
Katalog # APOSOR1029081-2.5K
Leverandør:  Apollo Scientific
New Product
CAS nummer:  25812-30-0
Gemfibrozil 98%

Bottle

CAS nummer:  25812-30-0
Price
9.040,00 kr
Log ind for tilgængelighed
1 * 2,5 kg
Product Details & Documents

Usage: Organic building block/ reagent.

Synonymer:  CI-719
  • CAS nummer:25812-30-0
  • Formel:C₁₅H₂₂O₃
  • Molecular Weight:250,33 g/mol
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer