Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Gemfibrozil 98%

Gemfibrozil 98%

Bottle
Katalog # APOSOR1029081-500G
New Product
Leverandør:   Apollo Scientific
Gemfibrozil 98%
Katalog # APOSOR1029081-500G
Leverandør:  Apollo Scientific
New Product
CAS nummer:  25812-30-0
Gemfibrozil 98%

Bottle

CAS nummer:  25812-30-0
Price
1.890,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents

Usage: Organic building block/ reagent.

Synonymer:  CI-719
  • CAS nummer:25812-30-0
  • Formel:C₁₅H₂₂O₃
  • Molecular Weight:250,33 g/mol
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.