Gemfibrozil 98%
Bottle
Katalog # APOSOR1029081-25G
New Product
Leverandør: Apollo Scientific
Gemfibrozil 98%
Katalog # APOSOR1029081-25G
Leverandør: Apollo Scientific
New Product
CAS nummer: 25812-30-0
Gå til:
Product Details & Documents
Usage: Organic building block/ reagent.
Synonymer: CI-719
- CAS nummer:25812-30-0
- Formel:C₁₅H₂₂O₃
- Molecular Weight:250,33 g/mol
Specifikationer
Specifikationer
- Pakning:Bottle



