Human Recombinant ERK1
Katalog # APOSBITP1400-5UG
New Product
Leverandør: Apollo Scientific
Human Recombinant ERK1
Katalog # APOSBITP1400-5UG
Leverandør: Apollo Scientific
New Product
CAS nummer:
Gå til:
Product Details & Documents
- Usage: Biochemical/life science applications
Specifikationer
Specifikationer
- Cat. No.:APOSBITP1400-5UG
- Enzym type:Recombinant
- Species:Human
- Enzym navn:ERK1
- Enzym synonymer:MAPK3 Inactive Enzyme;ERK1/MAPK3



