Orders LinkContactUsLinkComponent
D-(+)-Cellobiose
D-(+)-Cellobiose
Bottle
Katalog # APOSBIB6025-500G
Irriterende
New Product
Leverandør:   Apollo Scientific
D-(+)-Cellobiose
Katalog # APOSBIB6025-500G
Leverandør:  Apollo Scientific
New Product
CAS nummer:  528-50-7
D-(+)-Cellobiose
Bottle
CAS nummer:  528-50-7
Price
3.980,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents
Synonymer:  β-D-Glc-(1→4)-D-Glc, 4-β-D-glucopyranosyl-D-glueopyranose, Cellose, β-Cellobiose, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal, 4-O-β-D-Glucopyranosyl-D-glucopyranose
  • CAS nummer:528-50-7
  • Formel:C₁₂H₂₂O₁₁
  • MDL nummer:MFCD00136034
  • Molecular Weight:342.30 g/mol
  • Smeltende Pt:239…240 °C (dec.)
  • Opbevaringstemperatur:Stue temperatur
Documents
Specifikationer
Specifikationer
  • Pakning:
    Bottle
Specifikationer