Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Sodium Decanoate 98%

Bottle
Catalog # APOSOR1015017-5G
New Product
Supplier:   Apollo Scientific
Sodium Decanoate 98%
Catalog # APOSOR1015017-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  1002-62-6
Sodium Decanoate 98%

Bottle

CAS Number:  1002-62-6
Price
165,00 kr
Log in for availability
1 * 5 g
Product Details & Documents

Usage: Organic building block/reagent.

  • CAS Number:1002-62-6
  • Formula:C₁₀H₁₉NaO₂
  • MDL Number:MFCD00066453
  • Molecular Weight:194,25 g/mol
Specifications

Specifications

  • Pakning:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.