Orders LinkContactUsLinkComponent
SeramunBlock
SeramunBlock
SeramunBlock
Katalog # APOSBIL3122-5L
New Product
Leverandør:   Apollo Scientific
SeramunBlock
Katalog # APOSBIL3122-5L
Leverandør:  Apollo Scientific
New Product
CAS nummer:  
SeramunBlock
SeramunBlock
CAS nummer:  
Price
5.670,00 kr
Log ind for tilgængelighed
1 * 5 L
Product Details & Documents
Combined Stabiliser/ blocking solution for solid phase immobilised proteins effecting protein stabilisation and blocking of free surfaces.
Specifikationer
Specifikationer
  • Beskrivelse:
    SeramunBlock
Specifikationer