Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
ω-Brom-4-hydroxyacetophenon 97%

ω-Brom-4-hydroxyacetophenon 97%

Bottle
Katalog # APOSOR11266-10G
Ætsende
Irriterende
New Product
Leverandør:   Apollo Scientific
ω-Brom-4-hydroxyacetophenon 97%
Katalog # APOSOR11266-10G
Leverandør:  Apollo Scientific
New Product
CAS nummer:  2491-38-5
ω-Brom-4-hydroxyacetophenon 97%

Bottle

CAS nummer:  2491-38-5
Price
165,00 kr
Log ind for tilgængelighed
1 * 10 g
Product Details & Documents
Synonymer:  2-Bromo-1-(4-hydroxyphenyl)ethanone, 2-Bromo-4'-hydroxyacetophenone, 2-Bromo-1-(4-hydroxyphenyl)ethan-1-one, α-Bromo-4-hydroxyacetophenone, 4-Hydroxyphenacyl bromide
  • CAS nummer:2491-38-5
  • Formel:C₈H₇BrO₂
  • MDL nummer:MFCD00072424
  • Molecular Weight:215.05
  • Opbevaringstemperatur:Køleskab
  • FN:1759
  • ADR:8,III
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.