Orders LinkContactUsLinkComponent
(S)-(-)-Limonene 95%
(S)-(-)-Limonene 95%
Bottle
Katalog # APOSOR54353-25G
Brandfarlig
Miljø
Irriterende
New Product
Leverandør:   Apollo Scientific
(S)-(-)-Limonene 95%
Katalog # APOSOR54353-25G
Leverandør:  Apollo Scientific
New Product
CAS nummer:  5989-54-8
(S)-(-)-Limonene 95%
Bottle
CAS nummer:  5989-54-8
Price
253,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents
Usage: Organic building block/reagent
Synonymer:  (S)-(-)-p-Mentha-1,8-dien, (S)-(-)-Dipenten, (S)-(-)-1-Methyl-4-(1-methylvinyl)cyclohexen, L(-)-Limonen
  • CAS nummer:5989-54-8
  • Formel:C₁₀H₁₆
  • Molecular Weight:136,23404 g/mol
  • EINECS:227-815-6
  • FN:2052
Specifikationer
Specifikationer
  • Pakning:
    Bottle
Specifikationer