(S)-(-)-Limonene 95%
Bottle
Catalog # APOSOR54353-5G
New Product
Supplier: Apollo Scientific
(S)-(-)-Limonene 95%
Catalog # APOSOR54353-5G
Supplier: Apollo Scientific
New Product
CAS Number: 5989-54-8
Jump to:
Product Details & Documents
Usage: Organic building block/reagent
Synonyms: (S)-(-)-p-Mentha-1,8-dien, (S)-(-)-Dipenten, (S)-(-)-1-Methyl-4-(1-methylvinyl)cyclohexen, L(-)-Limonen
- CAS Number:5989-54-8
- Formula:C₁₀H₁₆
- Molecular Weight:136,23404 g/mol
- EINECS:227-815-6
- UN:2052
Specifications
Specifications
- Pakning:Bottle