(S)-(-)-Limonene 95%
Bottle
Katalog # APOSOR54353-1G
New Product
Leverandør: Apollo Scientific
(S)-(-)-Limonene 95%
Katalog # APOSOR54353-1G
Leverandør: Apollo Scientific
New Product
CAS nummer: 5989-54-8
Gå til:
Product Details & Documents
Usage: Organic building block/reagent
Synonymer: (S)-(-)-p-Mentha-1,8-dien, (S)-(-)-Dipenten, (S)-(-)-1-Methyl-4-(1-methylvinyl)cyclohexen, L(-)-Limonen
- CAS nummer:5989-54-8
- Formel:C₁₀H₁₆
- Molecular Weight:136,23404 g/mol
- EINECS:227-815-6
- FN:2052
Specifikationer
Specifikationer
- Pakning:Bottle



