Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Acetylthiocholiniodid

Acetylthiocholiniodid

Bottle
Catalog # APOSBIA4001-500G
Toxic
New Product
Supplier:   Apollo Scientific
Acetylthiocholiniodid
Catalog # APOSBIA4001-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  1866-15-5
Acetylthiocholiniodid

Bottle

CAS Number:  1866-15-5
Price
11.900,00 kr
Log in for availability
1 * 500 g
Product Details & Documents

Substrate for determination of acetylcholinesterase.

Synonyms:  (2-Mercaptoethyl)trimethylammonium iodide acetate, 2-(Acetylthio)-N,N,N-trimethylethan-1-aminium iodide, S-Acetylthiocholine iodide
  • CAS Number:1866-15-5
  • Formula:H₃CC(S)O(CH₂)₂N(CH₃)₃I
  • MDL Number:MFCD00011819
  • Molecular Weight:289.18 g/mol
  • Melting Pt:205…210 °C
  • Storage Temperature:Fryser
  • UN:2811
  • ADR:6.1,III

Documents

Specifications

Specifications

  • Pakning:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.