Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Acetylthiocholiniodid

Acetylthiocholiniodid

Bottle
Katalog # APOSBIA4001-100G
Giftig
New Product
Leverandør:   Apollo Scientific
Acetylthiocholiniodid
Katalog # APOSBIA4001-100G
Leverandør:  Apollo Scientific
New Product
CAS nummer:  1866-15-5
Acetylthiocholiniodid

Bottle

CAS nummer:  1866-15-5
Price
2.490,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

Substrate for determination of acetylcholinesterase.

Synonymer:  (2-Mercaptoethyl)trimethylammonium iodide acetate, 2-(Acetylthio)-N,N,N-trimethylethan-1-aminium iodide, S-Acetylthiocholine iodide
  • CAS nummer:1866-15-5
  • Formel:H₃CC(S)O(CH₂)₂N(CH₃)₃I
  • MDL nummer:MFCD00011819
  • Molecular Weight:289.18 g/mol
  • Smeltende Pt:205…210 °C
  • Opbevaringstemperatur:Fryser
  • FN:2811
  • ADR:6.1,III

Documents

Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.