Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Acetylthiocholiniodid

Acetylthiocholiniodid

Bottle
Katalog # APOSBIA4001-1G
Giftig
New Product
Leverandør:   Apollo Scientific
Acetylthiocholiniodid
Katalog # APOSBIA4001-1G
Leverandør:  Apollo Scientific
New Product
CAS nummer:  1866-15-5
Acetylthiocholiniodid

Bottle

CAS nummer:  1866-15-5
Price
187,00 kr
Log ind for tilgængelighed
1 * 1 g
Product Details & Documents

Substrate for determination of acetylcholinesterase.

Synonymer:  (2-Mercaptoethyl)trimethylammonium iodide acetate, 2-(Acetylthio)-N,N,N-trimethylethan-1-aminium iodide, S-Acetylthiocholine iodide
  • CAS nummer:1866-15-5
  • Formel:H₃CC(S)O(CH₂)₂N(CH₃)₃I
  • MDL nummer:MFCD00011819
  • Molecular Weight:289.18 g/mol
  • Smeltende Pt:205…210 °C
  • Opbevaringstemperatur:Fryser
  • FN:2811
  • ADR:6.1,III

Documents

Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer