Leptomycin B
Bottle
APOSBIL2101-50MG
New Product
Potent inhibitor of nuclear export.
Supplied as a 1mM solution in abs. ethanol
: (2E,5S,6R,7S,9R,10E,12E,15R,16Z,18E)-17-Ethyl-6-hydroxy-3,5,7,9,11,15-hexamethyl-19-((2S,3S)-3-methyl-6-oxo-3,6-dihydro-2H-pyran-2-yl)-8-oxononadeca-2,10,12,16,18-pentaenoic acid
- :87081-35-4
- :C₃₃H₄₈O₆
- :MFCD06795848
- :540.73 g/mol
- :11,1 °C
- :Fryser
- :1170
- :3,II
- Pakning:Bottle