Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Leptomycin B

Bottle
Katalog # APOSBIL2101-25MG
Brandfarlig
New Product
Leverandør:   Apollo Scientific
Leptomycin B
Katalog # APOSBIL2101-25MG
Leverandør:  Apollo Scientific
New Product
CAS nummer:  87081-35-4
Leptomycin B

Bottle

CAS nummer:  87081-35-4
Price
8.390,00 kr
Log ind for tilgængelighed
1 * 25 mg
Product Details & Documents

Potent inhibitor of nuclear export.

Supplied as a 1mM solution in abs. ethanol

Synonymer:  (2E,5S,6R,7S,9R,10E,12E,15R,16Z,18E)-17-Ethyl-6-hydroxy-3,5,7,9,11,15-hexamethyl-19-((2S,3S)-3-methyl-6-oxo-3,6-dihydro-2H-pyran-2-yl)-8-oxononadeca-2,10,12,16,18-pentaenoic acid
  • CAS nummer:87081-35-4
  • Formel:C₃₃H₄₈O₆
  • MDL nummer:MFCD06795848
  • Molecular Weight:540.73 g/mol
  • Flash Pt:11,1 °C
  • Opbevaringstemperatur:Fryser
  • FN:1170
  • ADR:3,II
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer