Brefeldin A
Bottle
Katalog # APOSBIB3042-250MG
New Product
Leverandør: Apollo Scientific
Brefeldin A
Katalog # APOSBIB3042-250MG
Leverandør: Apollo Scientific
New Product
CAS nummer: 20350-15-6
Gå til:
Product Details & Documents
BFA is a macrolide antiobiotic. Disrupts mechanism of Golgi apparatus.
Synonymer: (1R,2E,6S,10E,11aS,13S,14aR)-1,13-dihydroxy-6-methyl-6,7,8,9,12,13,14,14a-octahydro-1H-cyclopenta[f][1]oxacyclotridecin-4(11aH)-one, Brefeldin A, natural
- CAS nummer:20350-15-6
- Formel:C₁₆H₂₄O₄
- MDL nummer:MFCD00083258
- Molecular Weight:280.4 g/mol
- Smeltende Pt:200…205 °C
- Opbevaringstemperatur:Køleskab
- FN:2811
- ADR:6.1,III
Specifikationer
Specifikationer
- Pakning:Bottle
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



