Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Hesperidin

Hesperidin

Bottle
Katalog # APOSBIK9009-5G
New Product
Leverandør:   Apollo Scientific
Hesperidin
Katalog # APOSBIK9009-5G
Leverandør:  Apollo Scientific
New Product
CAS nummer:  520-26-3
Hesperidin

Bottle

CAS nummer:  520-26-3
Price
165,00 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents
Synonymer:  (S)-5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-((((2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-2-yl)oxy)chroman-4-one
  • CAS nummer:520-26-3
  • Formel:C₂₈H₃₄O₁₅
  • MDL nummer:MFCD00075663
  • Molecular Weight:610.56 g/mol
  • Smeltende Pt:250…255 °C
  • Opbevaringstemperatur:Køleskab
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.