Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Phthalsyre 98%

Phthalsyre 98%

Bottle
Catalog # APOSOR923932-2.5KG
New Product
Supplier:   Apollo Scientific
Phthalsyre 98%
Catalog # APOSOR923932-2.5KG
Supplier:  Apollo Scientific
New Product
CAS Number:  88-99-3
Phthalsyre 98%

Bottle

CAS Number:  88-99-3
Price
1.980,00 kr
Log in for availability
1 * 2,5 kg
Product Details & Documents
Synonyms:  Pathalic acid
  • CAS Number:88-99-3
  • Formula:C₈H₆O₄
  • MDL Number:MFCD00002467
  • Molecular Weight:166.13 g/mol
  • Storage Temperature:Stue temperatur
Specifications

Specifications

  • Pakning:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.