Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
4-Ethylanisol 98%

4-Ethylanisol 98%

Bottle
Katalog # APOSOR916875-500G
New Product
Leverandør:   Apollo Scientific
4-Ethylanisol 98%
Katalog # APOSOR916875-500G
Leverandør:  Apollo Scientific
New Product
CAS nummer:  1515-95-3
4-Ethylanisol 98%

Bottle

CAS nummer:  1515-95-3
Price
2.690,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents
Synonymer:  1-Ethyl-4-methoxybenzene
  • CAS nummer:1515-95-3
  • Formel:C₉H₁₂O
  • MDL nummer:MFCD00015176
  • Molecular Weight:136.19 g/mol
  • Opbevaringstemperatur:Stue temperatur
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.