Orders LinkContactUsLinkComponent
4-Ethylanisol 98%
4-Ethylanisol 98%
Bottle
  APOSOR916875-1G
New Product
4-Ethylanisol 98%
  APOSOR916875-1G
New Product
 :  1515-95-3
4-Ethylanisol 98%
Bottle
 :  1515-95-3
165,00 kr
1 * 1 g
 :  1-Ethyl-4-methoxybenzene
  •  :1515-95-3
  •  :C₉H₁₂O
  •  :MFCD00015176
  •  :136.19 g/mol
  •  :Stue temperatur
  • Pakning:
    Bottle