Lactic acid ≥95%
Bottle
Catalog # APOSBUP17254-500MG
New Product
Supplier: Apollo Scientific
Lactic acid ≥95%
Catalog # APOSBUP17254-500MG
Supplier: Apollo Scientific
New Product
CAS Number: 50-21-5
Gå til:
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonymer: DL-Mælkesyre
- CAS nummer:50-21-5
- Formel:C₃H₆O₃
- Molecular Weight:90,08 g/mol
- EINECS:200-018-0
- FN:3265
Specifikationer
Specifikationer
- Pakning:Bottle
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



