Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Lactic acid ≥95%

Lactic acid ≥95%

Bottle
Catalog # APOSBUP17254-100MG
Corrosive
New Product
Supplier:   Apollo Scientific
Lactic acid ≥95%
Catalog # APOSBUP17254-100MG
Supplier:  Apollo Scientific
New Product
CAS Number:  50-21-5
Lactic acid ≥95%

Bottle

CAS Number:  50-21-5
Price
396,00 kr
Log in for availability
1 * 100 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  DL-Mælkesyre
  • CAS Number:50-21-5
  • Formula:C₃H₆O₃
  • Molecular Weight:90,08 g/mol
  • EINECS:200-018-0
  • UN:3265
Specifications

Specifications

  • Pakning:
    Bottle

Specifications