Chloramphenicol ≥98%
Bottle
Katalog # APOSBUP18539-5G
New Product
Leverandør: Apollo Scientific
Chloramphenicol ≥98%
Katalog # APOSBUP18539-5G
Leverandør: Apollo Scientific
New Product
CAS nummer: 56-75-7
Gå til:
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonymer: Chloromycetin®, Tega-Cetin, (R,R)-Chloramphenicol
- CAS nummer:56-75-7
- Formel:C₁₁H₁₂Cl₂N₂O₅
- Molecular Weight:323,13 g/mol
Specifikationer
Specifikationer
- Pakning:Bottle



