Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Hydroxyamine hydrochloride ≥98%

Hydroxyamine hydrochloride ≥98%

Bottle
Catalog # APOSBUP18532-5G
New Product
Supplier:   Apollo Scientific
Hydroxyamine hydrochloride ≥98%
Catalog # APOSBUP18532-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  5470-11-1
Hydroxyamine hydrochloride ≥98%

Bottle

CAS Number:  5470-11-1
Price
955,00 kr
Log in for availability
1 * 5 g
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  Hydroxylamine HCl, Hydroxylamine hydrochloride
  • CAS Number:5470-11-1
  • Formula:ClH₄NO
  • Molecular Weight:69,49 g/mol
Specifications

Specifications

  • Pakning:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.