Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Hydroxyamine hydrochloride ≥98%

Hydroxyamine hydrochloride ≥98%

Bottle
Katalog # APOSBUP18532-1G
New Product
Leverandør:   Apollo Scientific
Hydroxyamine hydrochloride ≥98%
Katalog # APOSBUP18532-1G
Leverandør:  Apollo Scientific
New Product
CAS nummer:  5470-11-1
Hydroxyamine hydrochloride ≥98%

Bottle

CAS nummer:  5470-11-1
Price
428,00 kr
Log ind for tilgængelighed
1 * 1 g
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonymer:  Hydroxylamine HCl, Hydroxylamine hydrochloride
  • CAS nummer:5470-11-1
  • Formel:ClH₄NO
  • Molecular Weight:69,49 g/mol
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer