Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Fenofibrate ≥98%

Bottle
Katalog # APOSBUP17356-200MG
New Product
Leverandør:   Apollo Scientific
Fenofibrate ≥98%
Katalog # APOSBUP17356-200MG
Leverandør:  Apollo Scientific
New Product
CAS nummer:  49562-28-9
Fenofibrate ≥98%

Bottle

CAS nummer:  49562-28-9
Price
439,00 kr
Log ind for tilgængelighed
1 * 200 mg
Product Details & Documents

Usage: Biochemical/life science applications

Synonymer:  2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionic Acid Isopropyl Ester, Isopropyl 2-(4-(4-chlorobenzoyl)phenoxy)-2-methylpropanoate, Isopropyl 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionate
  • CAS nummer:49562-28-9
  • Formel:C₂₀H₂₁ClO₄
  • Molecular Weight:360,83 g/mol
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.