Finasteride ≥98%
Bottle
Catalog # APOSBUP14937-500MG
New Product
Supplier: Apollo Scientific
Finasteride ≥98%
Catalog # APOSBUP14937-500MG
Supplier: Apollo Scientific
New Product
CAS Number: 98319-26-7
Jump to:
Product Details & Documents
Usage: Biochemical/life science applications.
Synonyms: N-tert-Butyl-3-oxo-4-aza-5alpha-androst-1-en-17beta-carboxamide
- CAS Number:98319-26-7
- Formula:C₂₃H₃₆N₂O₂
- Molecular Weight:372,54 g/mol
Specifications
Specifications
- Pakning:Bottle



