Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Finasteride ≥98%

Finasteride ≥98%

Bottle
Katalog # APOSBUP14937-100MG
New Product
Leverandør:   Apollo Scientific
Finasteride ≥98%
Katalog # APOSBUP14937-100MG
Leverandør:  Apollo Scientific
New Product
CAS nummer:  98319-26-7
Finasteride ≥98%

Bottle

CAS nummer:  98319-26-7
Price
472,00 kr
Log ind for tilgængelighed
1 * 100 mg
Product Details & Documents

Usage: Biochemical/life science applications.

Synonymer:  N-tert-Butyl-3-oxo-4-aza-5alpha-androst-1-en-17beta-carboxamide
  • CAS nummer:98319-26-7
  • Formel:C₂₃H₃₆N₂O₂
  • Molecular Weight:372,54 g/mol
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.