Bacitracin
Bottle
Katalog # APOSBIB0106-100G
New Product
Leverandør: Apollo Scientific
Gå til:
Product Details & Documents
Bacitracin is active against gram-positive bacteria.
- Approx. 75,000 units/g
- CAS nummer:1405-87-4
- Formel:C₆₆H₁₀₃N₁₇O₁₆S₁
- MDL nummer:MFCD00062640
- Molecular Weight:1421.60 g/mol
- Smeltende Pt:221…225 °C
- Opbevaringstemperatur:Køleskab
Specifikationer
Specifikationer
- Pakning:Bottle



