Bacitracin
Bottle
Catalog # APOSBIB0106-50G
New Product
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Bacitracin is active against gram-positive bacteria.
- Approx. 75,000 units/g
- CAS Number:1405-87-4
- Formula:C₆₆H₁₀₃N₁₇O₁₆S₁
- MDL Number:MFCD00062640
- Molecular Weight:1421.60 g/mol
- Melting Pt:221…225 °C
- Storage Temperature:Køleskab
Specifications
Specifications
- Pakning:Bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



