Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Rosin 99,9%

Rosin 99,9%

Bottle
Katalog # APOSOR1026952-500G
New Product
Leverandør:   Apollo Scientific
Rosin 99,9%
Katalog # APOSOR1026952-500G
Leverandør:  Apollo Scientific
New Product
CAS nummer:  8050-09-7
Rosin 99,9%

Bottle

CAS nummer:  8050-09-7
Price
275,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents

Usage: Organic building block/ reagent.

Synonymer:  Terpentinoliefri harpiks
  • CAS nummer:8050-09-7
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.