Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Rosin 99,9%

Rosin 99,9%

Bottle
Catalog # APOSOR1026952-100G
New Product
Supplier:   Apollo Scientific
Rosin 99,9%
Catalog # APOSOR1026952-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  8050-09-7
Rosin 99,9%

Bottle

CAS Number:  8050-09-7
Price
165,00 kr
Log in for availability
1 * 100 g
Product Details & Documents

Usage: Organic building block/ reagent.

Synonyms:  Terpentinoliefri harpiks
  • CAS Number:8050-09-7
Specifications

Specifications

  • Pakning:
    Bottle

Specifications