Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Phthalic acid ≥98%

Phthalic acid ≥98%

Bottle
Katalog # APOSBUP19747-5G
New Product
Leverandør:   Apollo Scientific
Phthalic acid ≥98%
Katalog # APOSBUP19747-5G
Leverandør:  Apollo Scientific
New Product
CAS nummer:  88-99-3
Phthalic acid ≥98%

Bottle

CAS nummer:  88-99-3
Price
472,00 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents

Usage: Biochemical/life science applications.

Synonymer:  Pathalic acid
  • CAS nummer:88-99-3
  • Formel:C₈H₆O₄
  • Molecular Weight:166,13 g/mol
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer