D-(-)-Mandelic acid ≥95%
Bottle
Katalog # APOSBUP19814-10G
New Product
Leverandør: Apollo Scientific
D-(-)-Mandelic acid ≥95%
Katalog # APOSBUP19814-10G
Leverandør: Apollo Scientific
New Product
CAS nummer: 611-71-2
Gå til:
Product Details & Documents
Usage: Biochemical/life science applications.
Synonymer: D(-)-Mandelic acid, (R)-(-)-2-Hydroxy-2-phenylacetic acid, (R)-2-Hydroxy-2-phenylacetic acid, D(-)-2-Hydroxy-2-phenylacetic acid
- CAS nummer:611-71-2
- Formel:C₈H₈O₃
- Molecular Weight:152,15 g/mol
Specifikationer
Specifikationer
- Pakning:Bottle



