Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

DTNB ≥95%

Bottle
Catalog # APOSBUP19334-5G
Irritant
New Product
Supplier:   Apollo Scientific
DTNB ≥95%
Catalog # APOSBUP19334-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  69-78-3
DTNB ≥95%

Bottle

CAS Number:  69-78-3
Price
505,00 kr
Log in for availability
1 * 5 g
Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  DTNB, Ellman's reagent, 3,3'-Dithiobis(6-nitrobenzoic acid), 5,5'-Disulphanediylbis(2-nitrobenzoic acid), 2,2'-Dinitro-5,5'-dithiodibenzoic acid
  • CAS Number:69-78-3
  • Formula:C₁₄H₈N₂O₈S₂
  • Molecular Weight:396,35 g/mol
  • EINECS:200-714-4
Specifications

Specifications

  • Pakning:
    Bottle

Specifications

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.