DTNB ≥95%
Bottle
Katalog # APOSBUP19334-1G
New Product
Leverandør: Apollo Scientific
Gå til:
Product Details & Documents
Usage: Biochemical/life science applications.
Synonymer: DTNB, Ellman's reagent, 3,3'-Dithiobis(6-nitrobenzoic acid), 5,5'-Disulphanediylbis(2-nitrobenzoic acid), 2,2'-Dinitro-5,5'-dithiodibenzoic acid
- CAS nummer:69-78-3
- Formel:C₁₄H₈N₂O₈S₂
- Molecular Weight:396,35 g/mol
- EINECS:200-714-4
Specifikationer
Specifikationer
- Pakning:Bottle



