Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Leptomycin B ≥95%

Leptomycin B ≥95%

Bottle
Katalog # APOSBUP08666-50MG
New Product
Leverandør:   Apollo Scientific
Leptomycin B ≥95%
Katalog # APOSBUP08666-50MG
Leverandør:  Apollo Scientific
New Product
CAS nummer:  87081-35-4
Leptomycin B ≥95%

Bottle

CAS nummer:  87081-35-4
Price
18.020,00 kr
Log ind for tilgængelighed
1 * 50 mg
Product Details & Documents

Usage: Biochemical/life science applications.

Synonymer:  (2E,5S,6R,7S,9R,10E,12E,15R,16Z,18E)-17-Ethyl-6-hydroxy-3,5,7,9,11,15-hexamethyl-19-((2S,3S)-3-methyl-6-oxo-3,6-dihydro-2H-pyran-2-yl)-8-oxononadeca-2,10,12,16,18-pentaenoic acid
  • CAS nummer:87081-35-4
  • Formel:C₃₃H₄₈O₆
  • Molecular Weight:540,73 g/mol
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.