Leptomycin B ≥95%
Bottle
Catalog # APOSBUP08666-25MG
New Product
Supplier: Apollo Scientific
Leptomycin B ≥95%
Catalog # APOSBUP08666-25MG
Supplier: Apollo Scientific
New Product
CAS Number: 87081-35-4
Jump to:
Product Details & Documents
Usage: Biochemical/life science applications.
Synonyms: (2E,5S,6R,7S,9R,10E,12E,15R,16Z,18E)-17-Ethyl-6-hydroxy-3,5,7,9,11,15-hexamethyl-19-((2S,3S)-3-methyl-6-oxo-3,6-dihydro-2H-pyran-2-yl)-8-oxononadeca-2,10,12,16,18-pentaenoic acid
- CAS Number:87081-35-4
- Formula:C₃₃H₄₈O₆
- Molecular Weight:540,73 g/mol
Specifications
Specifications
- Pakning:Bottle
Specifications
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



